Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(-)-Mannitol

D-(-)-Mannitol

50 kg
Catalog # 0880-50KG
Supplier:   VWR Chemicals
D-(-)-Mannitol
Catalog # 0880-50KG
Supplier:  VWR Chemicals
CAS Number:  69-65-8
D-(-)-Mannitol
D-(-)-Mannitol

50 kg

CAS Number:  69-65-8
Price
1 520,00 €
Log in for availability
1 * 50 kg
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
Specifications

Specifications

  • Pk:
    50 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.