Eriochrome cyanine R, AnalaR NORMAPUR® analytical reagent C.I. 43820
Catalog # 86168.130
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Loss on drying (105°C):Max. 10,0 %
- Absorption Max. (buffer pH 7.0):434 --> 440 nm
- Spec.absorptivity(Lm;0.02g/l;pH 7.0;dry):130 --> 200
Specification Test Results
About this item
:
Storage Temperature:Ambient
EINECS:222-641-7
CAS Number:3564-18-9
Formula:C₂₃H₁₅Na₃O₉S
MDL Number:MFCD00012411