Order Entry
Austria
ContactUsLinkComponent
Peqlab peqGOLD Cycle-Pure™, PCR Clean-up, Kits
Peqlab peqGOLD Cycle-Pure™, PCR Clean-up, Kits
Supplier:  VWR Chemicals
Certificates
undefined
Peqlab peqGOLD Cycle-Pure™, PCR Clean-up, Kits
Supplier:  VWR Chemicals
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Sample size:
    5 to 50 µl enzymatic reactions
  • Description:
    Cycle-Pure Kit

Specifications

Related Information

Frequently Bought Together