Peqlab peqGOLD, DNA Isolation, Blood And Tissue DNA Mini Kits
Fast, easy, and economical kit for isolation of DNA from a variety of samples including fresh or frozen cultured cells, tissue, blood, buccal swabs and saliva.
- No need for organic extractions
- No need for time-consuming alcohol precipitations
- Enzymatic degradation of cellular RNA
- Efficient separation of enzyme inhibitors and contaminants
- High yields due to strong lysis
- Fast, purification in less than 20 min (after lysis)
Typical starting amount should be 30 mg tissue, 5×10⁶ cultured cells, or up to 250 µl. Gives purified DNA suitable for almost any downstream application.
: Kit contains peqGOLD DNA Mini columns, 2 ml collection tubes, BL buffer, TL buffer, Proteinase K solution, HBC buffer, DNA wash buffer and elution buffer.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:BacteriaBloodBiofluids (serum, plasma, saliva)Cultured cellsFFPE tissuesHuman/animal cells and tissues
- Target molecule:DNA
- Sample size:30 mg tissue, 5×10⁶ cultured cells, or up to 250 µl
- Description:Blood and tissue DNA mini kit
Frequently Bought Together