Water with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # HONCLC452-2.5
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle, GL45
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
Density:0.997 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:64-18-6
:7732-18-5