Order Entry
Austria
ContactUsLinkComponent
2-Propanol ≥99.8% VLSI, Semiconductor grade (Honeywell 17604)
Flammable
Irritant
2-Propanol ≥99.8% VLSI, Semiconductor grade (Honeywell 17604)
Catalog # HONC40301H-2.5L
CAS Number:  
Flammable
Irritant
2-Propanol ≥99.8% VLSI, Semiconductor grade (Honeywell 17604)
Catalog # HONC40301H-2.5L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Particles > 0,5 µm:
    Max. 250 P/ml
  • Assay (GC):
    Min. 99.8 %
  • Boiling range:
    81 - 83 °C
  • Density (D 20/4):
    0.785 - 0.787
  • Refractive index (n 20/D):
    1.3770 - 1.3780
  • Non-volatile matter:
    Max. 5 ppm
  • Water (Karl Fischer):
    Max. 500 ppm
  • Free acid (as C₂H₅COOH):
    Max. 10 ppm
  • Silver (Ag):
    Max. 0.01 ppm
  • Aluminium (Al):
    Max. 0.05 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Gold (Au):
    Max. 0.02 ppm
  • Boron (B):
    Max. 0.01 ppm
  • Barium (Ba):
    Max. 0.02 ppm
  • Beryllium (Be):
    Max. 0.01 ppm
  • Bismuth (Bi):
    Max. 0.02 ppm
  • Calcium (Ca):
    Max. 0.05 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.05 ppm
  • Gallium (Ga):
    Max. 0.01 ppm
  • Indium (In):
    Max. 0.01 ppm
  • Potassium (K):
    Max. 0.02 ppm
  • Lithium (Li):
    Max. 0.01 ppm
  • Magnesium (Mg):
    Max. 0.02 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.01 ppm
  • Sodium (Na):
    Max. 0.1 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Platinum (Pt):
    Max. 0.02 ppm
  • Antimony (Sb):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.02 ppm
  • Strontium (Sr):
    Max. 0.01 ppm
  • Titanium (Ti):
    Max. 0.01 ppm
  • Thallium (Tl):
    Max. 0.01 ppm
  • Vanadium (V):
    Max. 0.01 ppm
  • Zinc (Zn):
    Max. 0.02 ppm
  • Zirconium (Zr):
    Max. 0.01 ppm
  • Chloride (Cl):
    Max. 1 ppm
  • Sulfate (SO₄):
    Max. 1 ppm
  • Acetone (GC):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :60.1 g/mol
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-8
UN:1219
ADR:3,II
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011675