Order Entry
Austria
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™
N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™
Certificates
undefined
N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Non-volatile matter:
    Max. 0.001 %
  • Amines (as dimethylamine):
    Max. 0.0002 %

Specification Test Results

Related Information

Frequently Bought Together