Order Entry
Austria
ContactUsLinkComponent
 
Tetrahydrofuran ≥99.5% (by GC), AR®, Macron Fine Chemicals™
 
undefined
Tetrahydrofuran ≥99.5% (by GC), AR®, Macron Fine Chemicals™
 

 

  • Environmentally Preferable:

 

 

  • Assay (GC):
    ≥99.5%
  • Water (KF:
    ≤0.05 %
  • Titrable Acid (meq/g):
    ≤0.0005
  • Residue after Evaporation:
    ≤0.0005 %
  • Peroxides (as H₂O₂):
    ≤0.015 %

 

Frequently Bought Together