Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
 
Cyclohexane ≥99.5% (by GC), ChromAR® for HPLC, Macron Fine Chemicals™
  6720-25
 :  
undefined
 
 
 
 
Cyclohexane ≥99.5% (by GC), ChromAR® for HPLC, Macron Fine Chemicals™
  6720-25
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Assay (GC):
    ≥99.5%
  • Residue after Evaporation:
    ≤0.0005 %
  • Titrable Acid (meq/g):
    ≤0.0005
  • Water (KF:
    ≤0.01 %
  • 210 nm:
    ≤0.4
  • 220 nm:
    ≤0.1
  • 230 nm:
    ≤0.05
  • 240 nm:
    ≤0.01
  • 350 nm:
    ≤0.004
  • undefined:
    .

 

 

 :
 :84.16 g/mol
 :–18 °C
 :Ambient
 :1145
 :3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
 :110-82-7
 :C₆H₁₂
 :MFCD00003814