ContactUsLinkComponent
Avantor® Hichrom, Frit Removal Tool
Avantor® Hichrom, Frit Removal Tool
Catalog # HI-300
Supplier:  Avantor
undefined
Avantor® Hichrom, Frit Removal Tool
Catalog # HI-300
Supplier:  Avantor
Specifications
  • For:
    HPLC/LC columns
  • Description:
    Frit removal tool
  • Pk:
    1
Specifications
About this item
The frit removal tool is ideal for the removal of frits used in HPLC/LC columns.