ContactUsLinkComponent
Avantor® Hichrom PEEKsil™, Tubing
Catalog # HICHHI-1083
Supplier:  Avantor
undefined
Avantor® Hichrom PEEKsil™, Tubing
Catalog # HICHHI-1083
Supplier:  Avantor
Specifications
  • Description:
    PEEKsil™ tubing for HPLC, LC/MS applications
  • Length:
    500 mm
  • Seq:
    02
  • Pk:
    2
Specifications
About this item
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility.