ContactUsLinkComponent
N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
  D-074-1ML
 :  Merck
 :  
undefined
N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
  D-074-1ML
 :  Merck
 :  
  • Concentration:
    100 µg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    N-Desmethylcitalopram-d3 hydrochloride solution
  • Description:
    N-Desmethylcitalopram-d3 hydrochloride solution
  • Pk Info:
    Ampoule of 1 ml
A stable-labeled internal standard of N-Desmethylcitalopram, an active metabolite of the SSRI antidepressant drug citalopram.
 : Snap-N-Spike®/Snap-N-Shoot®