ContactUsLinkComponent
Zorbax CN, HPLC Columns
Zorbax CN, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Zorbax CN, HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • ZCN-150A
  • ZCN-250A
  • ZCN-250AF
  • ZCN-150AS
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 7%
  • 7%
  • 7%
  • 7%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • 330 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)