ContactUsLinkComponent
Laboratory coats, SMS, with zip closure
Laboratory coats, SMS, with zip closure
Certificates
undefined
Laboratory coats, SMS, with zip closure
Specifications
  • Catalog No:
  • HOIT120841
  • HOIT114520
  • HOIT114521
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Laboratory
  • Laboratory
  • Laboratory
  • Weight:
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²