Order Entry
Austria
ContactUsLinkComponent
Programmed test mix, in dichloromethane (varied conc.), certified reference material, Supelco®
Programmed test mix, in dichloromethane (varied conc.), certified reference material, Supelco®
Catalog # 47304.
Supplier:  Merck
CAS Number:  
Programmed test mix, in dichloromethane (varied conc.), certified reference material, Supelco®
Catalog # 47304.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Programmed Test Mix, in dichloromethane (varied conc.)
  • Storage temperature:
    2 - 8 °C
  • Pk Info:
    ampule of 2 mL

Specifications

About this item

This test mix is based on the comprehensive mix developed by Grob (Grob, et al., J. Chromatogr. 156 (1978) p. 1)

Analytes:

2,3-Butanediol 530 μg/ml; Decane 280 μg/ml; Dicyclohexylamine 310 μg/ml; 2,6-Dimethylaniline 320 μg/ml; 2,6-Dimethylphenol 320 μg/ml; 2-Ethylhexanoic acid 380 μg/ml; Methyl decanoate 420 μg/ml; Methyl laurate 410 μg/ml; Methyl undecanoate 420 μg/ml; Nonanal 400 μg/ml; 1-Octanol 360 μg/ml; Undecane 290 μg/ml

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.