ContactUsLinkComponent
IQ/OQ/PQ Kit for Ion Chromatography, certified reference material, Supelco®
IQ/OQ/PQ Kit for Ion Chromatography, certified reference material, Supelco®
Catalog # 12674-1KT
Supplier:  Merck
CAS Number:  
IQ/OQ/PQ Kit for Ion Chromatography, certified reference material, Supelco®
Catalog # 12674-1KT
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Standards:
    Certified reference material
  • Description:
    IQ/OQ/PQ Kit for ion chromatography
  • M-Clarity quality level:
    MQ300
Specifications
About this item
IQ/OQ/PQ-Kit for the qualification of ion chromatography systems. Suitable for the verification of linearity, repeatability and carry-over.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034
Packaging: Contains six certified bromide standards (5, 10, 20, 50, 100 and 1000 mg/kg; 100 ml each). The exact contents, traceability and uncertainty are given in a detailed certificate.