ContactUsLinkComponent
HPLC isocratic systems diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
HPLC isocratic systems diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Catalog # 48270-U
Supplier:  Merck
CAS Number:  
HPLC isocratic systems diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Catalog # 48270-U
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    HPLC Isocratic Systems Diagnostics Mix, in methanol: water (3:2) (varied conc.)
  • M-Clarity quality level:
    MQ100
  • Pk Info:
    pkg of 6 × 1 ml
Specifications
About this item
This formulation is designed for evaluating how reliably a chromatographic system is providing such fundamentally important parameters as flow rate, proportioning, and mixing.
Analytes:
Butylparaben 25 μg/ml; Ethyl 4-hydroxybenzoate 14 μg/ml; Methyl 4-hydroxybenzoate 10 μg/ml; Propyl 4-hydroxybenzoate 15 μg/ml
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.