Order Entry
Austria
ContactUsLinkComponent
Methylated DNA capture kit
Methylated DNA capture kit
Catalog # ABNOKA0691
Supplier:  Abnova
CAS Number:  
Methylated DNA capture kit
Catalog # ABNOKA0691
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Methylated DNA Capture Kit

Specifications

About this item

Methylated DNA Capture Kit is used to enrich methylated DNA.

Storage Instructions: Store the following components at 4 °C: MC1, MC3, normal mouse IgG, anti-5-methylcytosine, proteinase K, 8 well strips. Store other components at room temperature.

Caution: For research use only (RUO)