Order Entry
Austria
ContactUsLinkComponent
 
Sodium thiosulphate solution 0.01 mol/l (0.01 N)), Titripur®, Supelco®
Catalog # 1.60318.1003
Supplier:  Merck
CAS Number:  
Sodium thiosulphate solution 0.01 mol/l (0.01 N)), Titripur®, Supelco®
Catalog # 1.60318.1003
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle with RFID tag
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :158.11 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499