Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Supelco® Organic Reference Standard, Testosterone solution

Testosterone solution, 1,0 mg/ml in 1,2-dimethoxyethane, Drug standard, 1 mL
Catalog # T5411-1ML
Supplier:   Merck
Supelco® Organic Reference Standard, Testosterone solution
Catalog # T5411-1ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Testosterone solution
Supelco® Organic Reference Standard, Testosterone solution

Testosterone solution, 1,0 mg/ml in 1,2-dimethoxyethane, Drug standard, 1 mL

CAS Number:  
Price
103,00 €
Log in for availability
1 * 1 mL
Product Details & Documents
  • GHS signal: GHS02, GHS08, GHS07

 : P202 - P210 - P233 - P303 + P361 + P353 - P304 + P340 + P312 - P308 + P313

Specifications

Specifications

  • Concentration:
    1,0 mg/ml in 1,2-dimethoxyethane
  • Pk:
    1 mL
  • Standards:
    Drug standard
  • Description:
    Testosterone solution
  • Cat. No.:
    T5411-1ML
  • M-Clarity quality level:
    MQ200
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H315 - H332 - H360FD
  • MDL No.:
    MFCD00003654
  • Flash point:
    5 °C - Closed cup
  • CAS No.:
    58-22-0
  • Shipping conditions:
    STD

Specifications