Supelco® Organic Reference Standard, Digoxigenin
Supplier: Merck
Certificates
About this item
- GHS signal: GHS06
Caution: P260 - P262 - P264 - P280 - P302 + P352 + P310 - P304 + P340 + P310
Specifications
- CAS No.:1672-46-4
- Storage conditions:RT
- Hazard class description:H300 + H310 + H330
- Shipping conditions:STD
- MDL No.:MFCD00871861
- M-Clarity quality level:MQ100
- Synonym(s):3β,12β,14β,21-Tetrahydroxy-20(22)-norcholenic acid lactone; 3β,12β,14-Trihydroxy-5β,20(22)-cardenolide; 5β,20(22)-Cardenolide-3β,12β,14-triol; Lanadigigenin
- Melting point:222 °C (lit.)
- Molecular weight:390,51
Specifications
Frequently Bought Together