Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
O-Desmethylnaproxen, analytical standard, Supelco®
O-Desmethylnaproxen, analytical standard, Supelco®
O-Desmethylnaproxen, analytical standard, Supelco®
95114-25MG
:
Merck
:
O-Desmethylnaproxen, analytical standard, Supelco®
95114-25MG
:
Merck
:
Pk:
25 mg
Standards:
analytical standard
CAS No.:
52079-10-4
Description:
O-Desmethylnaproxen
Description:
O-Desmethylnaproxen