Order Entry
Austria
ContactUsLinkComponent
 
Melting point standards, potassium nitrate, traceable to primary standards, Supelco®
Catalog # 49143-5G
Supplier:  Merck
CAS Number:  
undefined
Melting point standards, potassium nitrate, traceable to primary standards, Supelco®
Catalog # 49143-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Melting pt.:
    334,0 °C
  • Accuracy:
    ± 0,3 °C

Specifications

About this item

Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC).