ContactUsLinkComponent
Cleanroom mop, Clino One Way Ultra PW-I
Cleanroom mop, Clino One Way Ultra PW-I
Catalog # 141-1457
undefined
Cleanroom mop, Clino One Way Ultra PW-I
Catalog # 141-1457
Specifications
  • Colour:
    White
  • Description:
    Clino One Way Ultra PW-I mop, 55 g
  • L×W:
    400×120 mm
  • Pk:
    5
Specifications
About this item
Pre-washed and gamma-irradiated sterile premium disposable mop with maximum square meter output.
  • For use on all standard cleanroom and pharmaceutical floors
  • 100% Polyester microfiber material
  • With backfilling for enormous sqm performance
  • Produced in a controlled environment, gamma irradiated
Excellent cleaning performance due to polyester microfibre with special structure for optimum particle absorption. Up to 20 m² area output due to back-feeding. Very high-quality processing suitable for cleanrooms ISO 5, 6, 7, 8, GMP A/B, C, D.