Order Entry
Austria
ContactUsLinkComponent
Fluoride ion solution for ISE, Supelco®, Merck
Fluoride ion solution for ISE, Supelco®, Merck
Catalog # 47072-100ML
Supplier:  Merck
CAS Number:  
Fluoride ion solution for ISE, Supelco®, Merck
Catalog # 47072-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0.1 M
  • Pk:
    100 mL
  • Description:
    Sodium fluoride solution

Specifications

About this item