Sucrose standard for IC, TraceCERT®, Supelco®
Catalog # 69631-50ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/l
- Pk:50 mL
- CAS No.:57-50-1
- Description:Sucrose in water
- Formula:C₁₂H₂₂O₁₁
- Description:Sucrose
Specifications
About this item
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.