Succinate standard for IC, TraceCERT®, Supelco®
Artikelnr. 43057-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/l
- Pk:100 mL
- Description:Succinate in water
Specifications
About this item
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.