Order Entry
Austria
ContactUsLinkComponent
Ammonium Nitrogen standard for IC, TraceCERT®, Supelco®
Ammonium Nitrogen standard for IC, TraceCERT®, Supelco®
Catalog # 89503-100ML
Supplier:  Merck
CAS Number:  
undefined
Ammonium Nitrogen standard for IC, TraceCERT®, Supelco®
Catalog # 89503-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Description:
    Ammonium standard solution
  • Element:
    Nitrogen (as NH₄+)

Specifications

About this item

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.