Order Entry
Austria
ContactUsLinkComponent
Tungsten Standard for ICP, TraceCERT®, Supelco®
Tungsten Standard for ICP, TraceCERT®, Supelco®
Catalog # 50334-100ML
Supplier:  Merck
CAS Number:  
undefined
Tungsten Standard for ICP, TraceCERT®, Supelco®
Catalog # 50334-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃/2% HF
  • Pk:
    100 mL
  • Element:
    Tungsten
  • Symbol:
    W

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034