Order Entry
Austria
ContactUsLinkComponent
Thallium Standard for ICP, TraceCERT®, Supelco®
Thallium Standard for ICP, TraceCERT®, Supelco®
Catalog # 05295-100ML
Supplier:  Merck
CAS Number:  
Thallium Standard for ICP, TraceCERT®, Supelco®
Catalog # 05295-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Element:
    Thallium
  • Symbol:
    Tl

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034