Order Entry
Austria
ContactUsLinkComponent
 
Tellurium Standard for ICP, TraceCERT®, Supelco®
  78358-100ML
 :  Merck
 :  
Tellurium Standard for ICP, TraceCERT®, Supelco®
  78358-100ML
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HNO₃/tr HF
  • Pk:
    100 mL
  • Element:
    Tellurium
  • Symbol:
    Te

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034