Order Entry
Austria
Orders LinkContactUsLinkComponent
Selenium Standard for ICP, TraceCERT®, Supelco®

Selenium Standard for ICP, TraceCERT®, Supelco®

Selenium, Se, 10000 mg/L, Nitric acid dil., 100 mL
Catalog # 98838-100ML
Supplier:   Merck
Selenium Standard for ICP, TraceCERT®, Supelco®
Catalog # 98838-100ML
Supplier:  Merck
CAS Number:  
Selenium Standard for ICP, TraceCERT®, Supelco®
Selenium Standard for ICP, TraceCERT®, Supelco®

Selenium, Se, 10000 mg/L, Nitric acid dil., 100 mL

CAS Number:  
Price
375,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034

Specifications

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.