Rhenium Standard for ICP, TraceCERT®, Supelco®
Catalog # 39957-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pk:100 mL
- Element:Rhenium
- Symbol:Re
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034