Order Entry
Austria
ContactUsLinkComponent
Palladium atomic absorption standard solution, analytical standard, Supelco®
Palladium atomic absorption standard solution, analytical standard, Supelco®
Catalog # 207349-500ML
Supplier:  Merck
CAS Number:  
Palladium atomic absorption standard solution, analytical standard, Supelco®
Catalog # 207349-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HCl
  • Pk:
    500 mL
  • Element:
    Palladium
  • Symbol:
    Pd

Specifications

About this item

Exact Pd concentration on label.