Order Entry
Austria
ContactUsLinkComponent
Multi-element standard solution 4 for ICP, TraceCERT®, Supelco®
Multi-element standard solution 4 for ICP, TraceCERT®, Supelco®
Catalog # 51844-100ML
Supplier:  Merck
CAS Number:  
Multi-element standard solution 4 for ICP, TraceCERT®, Supelco®
Catalog # 51844-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Matrix:
    10% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi-element standard solution 4 for ICP
  • M-Clarity quality level:
    MQ300

Specifications

About this item

17 components

Al, 40 mg/l; As, 40 mg/l; B, 100 mg/l; Ba, 40 mg/l; Be, 10 mg/l
Cd, 10 mg/l; Co, 10 mg/l; Cr, 20 mg/l; Cu, 20 mg/l; Fe, 100 mg/l
Mn, 10 mg/l; Ni, 20 mg/l; Pb, 40 mg/l; Se, 100 mg/l; Tl, 100 mg/l
V, 40 mg/l; Zn, 100 mg/l

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034