Order Entry
Austria
ContactUsLinkComponent
Multi-element standard solution 3 for ICP, TraceCERT®, Supelco®
Multi-element standard solution 3 for ICP, TraceCERT®, Supelco®
Catalog # 49596-100ML
Supplier:  Merck
CAS Number:  
undefined
Multi-element standard solution 3 for ICP, TraceCERT®, Supelco®
Catalog # 49596-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi-element standard solution 3 for ICP
  • M-Clarity quality level:
    MQ300

Specifications

About this item

Ca, 2000 mg/l
K, 200 mg/l
Mg, 400 mg/l
Na, 1000 mg/l

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034