Order Entry
Austria
ContactUsLinkComponent
 
Multi-cation standard 2 for IC, TraceCERT®, Supelco®
Catalog # 93159-100ML
Supplier:  Merck
CAS Number:  
undefined
Multi-cation standard 2 for IC, TraceCERT®, Supelco®
Catalog # 93159-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Description:
    Multi Cation Standard 2 for IC

Specifications

About this item

Ba, 100 mg/l; Ca, 100 mg/l; K, 100 mg/l
Li, 100 mg/l; Mg, 100 mg/l; Mn, 100 mg/l
Na, 100 mg/l; NH4, 100 mg/l; Sr, 100 mg/l

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034