Order Entry
Austria
ContactUsLinkComponent
Multi cation standard 1 for IC, TraceCERT®, Supelco®
Multi cation standard 1 for IC, TraceCERT®, Supelco®
Catalog # 91286-100ML
Supplier:  Merck
CAS Number:  
undefined
Multi cation standard 1 for IC, TraceCERT®, Supelco®
Catalog # 91286-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Matrix:
    0,1% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi Cation Standard 1 for IC

Specifications

About this item

Ca, 1000 mg/l
K, 200 mg/l
Li, 50 mg/l
Mg, 200 mg/l
Na, 200 mg/l
NH4, 400 mg/l

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034