Order Entry
Austria
ContactUsLinkComponent
Lithium standard for IC, TraceCERT®, Supelco®
Lithium standard for IC, TraceCERT®, Supelco®
Catalog # 59878-100ML
Supplier:  Merck
CAS Number:  
undefined
Lithium standard for IC, TraceCERT®, Supelco®
Catalog # 59878-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034