Order Entry
Austria
ContactUsLinkComponent
Europium atomic absorption standard solution, analytical standard, Supelco®
Europium atomic absorption standard solution, analytical standard, Supelco®
Catalog # 207128-100ML
Supplier:  Merck
CAS Number:  
Europium atomic absorption standard solution, analytical standard, Supelco®
Catalog # 207128-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Europium
  • Symbol:
    Eu

Specifications

About this item