Copper Standard for IC, TraceCERT®, Supelco®
Catalog # 40786-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pk:100 mL
- M-Clarity quality level:MQ300
- Element:Copper
- Symbol:Cu
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034