Order Entry
Austria
ContactUsLinkComponent
Cadmium standard for ICP-MS, TraceCERT®, Supelco®
Cadmium standard for ICP-MS, TraceCERT®, Supelco®
Catalog # 12313-100ML
Supplier:  Merck
CAS Number:  
Cadmium standard for ICP-MS, TraceCERT®, Supelco®
Catalog # 12313-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034