Order Entry
Austria
ContactUsLinkComponent
Phloxin B for histology, for cytology
Phloxin B for histology, for cytology
Catalog # 720-1159
Supplier:  RAL
CAS Number:  
Phloxin B for histology, for cytology
Catalog # 720-1159
Supplier:  RAL
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :829.6344 g/mol
EINECS: 242-355-6
 :1
CAS Number:18472-87-2
Formula:C₂₀H₂Br₄Cl₄Na₂O₅
MDL Number:MFCD00070627