Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide solution 0.1 mol/l (0.1 N) in 2-propanol acc. to DIN 51558 part 1, AVS TITRINORM
Potassium hydroxide solution 0.1 mol/l (0.1 N) in 2-propanol acc. to DIN 51558 part 1, AVS TITRINORM
Catalog # 86190.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydroxide solution 0.1 mol/l (0.1 N) in 2-propanol acc. to DIN 51558 part 1, AVS TITRINORM
Catalog # 86190.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553