Order Entry
Austria
ContactUsLinkComponent
Copper(II) sulphate solution 0.1 mol/l (0.1 M), AVS TITRINORM
Environment
Copper(II) sulphate solution 0.1 mol/l (0.1 M), AVS TITRINORM
Catalog # 86183.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Environment
Copper(II) sulphate solution 0.1 mol/l (0.1 M), AVS TITRINORM
Catalog # 86183.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :159.61 g/mol
Storage Temperature:Ambient
UN:3082
ADR:9,III
 :GHS09
CAS Number:7758-98-7
Formula:CuSO₄
MDL Number:MFCD00010981