

Nitric acid 10 mol/l (10 N), AVS TITRINORM
Catalog # 86172.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Titer (20°C):0.9980 – 1.0020
- Method of analysis:visual titration
- Using:sodium hydroxide volumetric solution
- Indicator:methyl red
- Reference material:indirect potassium hydrogenphthalate (KHP)
- Traceability:the reference material is a secondary reference material, traceable to standard material from NIST
Specification Test Results
About this item
:
:63.01 g/mol
UN:2031
ADR:8,II
:GHS05
:GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349