Order Entry
Austria
ContactUsLinkComponent
Nerve growth factor receptor (NGFR/P75ECD) Rapid™ ELISA kit: Human
Nerve growth factor receptor (NGFR/P75ECD) Rapid™ ELISA kit: Human
Supplier:  Biosensis
Certificates
undefined
Nerve growth factor receptor (NGFR/P75ECD) Rapid™ ELISA kit: Human
Supplier:  Biosensis

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Cross reactivity:
    The antibodies used in this ELISA kit does not cross-react with mouse p75ECD protein
  • Specificity:
    Human
  • Detection range:
    62,5 - 4000 pg/ml
  • Sensitivity:
    < 50 pg/ml

Specifications

Frequently Bought Together