Picrofuchsin solution acc. to van Gieson used in nuclear staining, for histology, Sigma-Aldrich®
Catalog # 1.00199.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 ml
- Seq:2725
- Pk Info:Preparation Capacity 500 slides
Specifications
Specification Test Results
- Suitability for microscopy:passes test
- Nuclei:black-brown
- Collagen:red
- Cytoplasm:yellow
- musculature (muscles):yellow
- colloid:yellow to red
- glia fibrils:yellow
- Mucus:yellow to red
- hyaline:yellow to red
- Amyloid:yellow to red
- cornified epithelium:yellow to red
Specification Test Results
About this item
:
Boiling Pt:100 °C
Storage Temperature:Ambient
:1