ContactUsLinkComponent
Papanicolaou's solution 3d polychromatic solution EA 65 used in nuclear staining, for cytology, Sigma-Aldrich®
Flammable
Irritant
Papanicolaou's solution 3d polychromatic solution EA 65 used in nuclear staining, for cytology, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
Papanicolaou's solution 3d polychromatic solution EA 65 used in nuclear staining, for cytology, Sigma-Aldrich®
Supplier:  Merck
Specification Test Results
  • Suitability for microscopy (Vaginal smear):
    passes test
  • Nuclei:
    blue to dark violet
  • Cyanophilic cytoplasm:
    blue-green to blue-grey
  • Eosinophilic cytoplasm:
    pink to red-violet