Order Entry
Austria
ContactUsLinkComponent
Shorr staining solution stain for histology, Sigma-Aldrich®
Shorr staining solution stain for histology, Sigma-Aldrich®
Catalog # 1.09275.0500
Supplier:  Merck
CAS Number:  
Shorr staining solution stain for histology, Sigma-Aldrich®
Catalog # 1.09275.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Suitability for microscopy (Vaginal smear):
    passes test
  • Nuclei:
    brownish red
  • Eosinophilic cytoplasm:
    red
  • Cyanophilic cytoplasm:
    blue-green

Specification Test Results

About this item

 :
ADR:3,II
 :M-Clarity™ quality level = MQ300
UN:1993
 :GHS02
 :GHS07
Storage Temperature:Ambient
 :1